Assay Details
| Gene Symbol: ENO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005652595.3 | XP_005652652.1 | 1085 | 64 | ||
| XM_021092412.1 | XP_020948071.1 | 659 | 64 | |||
| GenBank mRNA | AK237968.1 | - | 8 - 9 | 998 | 64 | |
| AK391707.1 | - | 8 - 9 | 935 | 64 | ||
| AK392135.1 | - | 8 - 9 | 949 | 64 | ||
More Information
Set Membership: |
|

ENO2
Genomic Map