Assay Details
| Gene Symbol: MIA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005659906.3 | XP_005659963.1 | 2224 | 68 | ||
| XM_005659907.3 | XP_005659964.1 | 1604 | 68 | |||
| XM_005659909.3 | XP_005659966.1 | 2305 | 68 | |||
| XM_021073967.1 | XP_020929626.1 | 3582 | 68 | |||
| XM_021073969.1 | XP_020929628.1 | 3567 | 68 | |||
| XM_021074004.1 | XP_020929663.1 | 1649 | 68 | |||
| XM_021074012.1 | XP_020929671.1 | 1737 | 68 | |||
| XM_021074016.1 | XP_020929675.1 | 1614 | 68 | |||
| GenBank mRNA | AK237997.1 | - | 7 - 8 | 582 | 68 | |
More Information
Set Membership: |
|

MIA2
Genomic Map