Assay Details
| Gene Symbol: MCIDAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021077052.1 | XP_020932711.1 | 1775 | 77 | ||
| GenBank mRNA | AK238080.1 | - | 1 - 2 | 133 | 77 | |
More Information
Set Membership: |
|

MCIDAS
Genomic Map