Assay Details
| Gene Symbol: TMPRSS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005653708.3 | XP_005653765.1 | 825 | 58 | ||
| XM_013977479.2 | XP_013832933.1 | 759 | 58 | |||
| XM_021062909.1 | XP_020918568.1 | 999 | 58 | |||
| XM_021062910.1 | XP_020918569.1 | 790 | 58 | |||
| GenBank mRNA | AK238098.1 | - | 5 - 6 | 676 | 58 | |
| AK349918.1 | - | 5 - 6 | 455 | 58 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021062910.1 transcripts.
Set Membership: |
|

TMPRSS4
Genomic Map