Assay Details
| Gene Symbol: SRSF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003125210.5 | XP_003125258.1 | 4769 | 93 | ||
| XM_003125211.5 | XP_003125259.1 | 4769 | 93 | |||
| XM_005662599.3 | XP_005662656.1 | 4769 | 93 | |||
| XM_005662601.3 | XP_005662658.1 | 4769 | 93 | |||
| XM_021087603.1 | XP_020943262.1 | 4769 | 93 | |||
| XM_021087604.1 | XP_020943263.1 | 4769 | 93 | |||
| GenBank mRNA | AK238144.1 | - | 3 - 4 | 490 | 93 | |
| AK391744.1 | - | 3 - 4 | 533 | 93 | ||
More Information
Set Membership: |
|

SRSF7
Genomic Map