Assay Details
| Gene Symbol: AK8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021070821.1 | XP_020926480.1 | 959 | 67 | ||
| XM_021070826.1 | XP_020926485.1 | 1034 | 67 | |||
| XM_021070832.1 | XP_020926491.1 | 958 | 67 | |||
| XM_021070835.1 | XP_020926494.1 | 966 | 67 | |||
| GenBank mRNA | AK238275.1 | - | 9 - 10 | 903 | 67 | |
| AK240567.1 | - | 9 - 10 | 938 | 67 | ||
| AK399493.1 | - | 9 - 10 | 958 | 67 | ||
More Information
Set Membership: |
|

AK8
Genomic Map