Assay Details
| Gene Symbol: BABAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021088348.1 | XP_020944007.1 | 287 | 158 | ||
| XM_021088349.1 | XP_020944008.1 | 287 | 158 | |||
| GenBank mRNA | AK238300.1 | - | 2 - 3 | 226 | 158 | |
More Information
Set Membership: |
|

BABAM2
Genomic Map