Assay Details
| Gene Symbol: LOC100624460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021085118.1 | XP_020940777.1 | 19081 | 72 | ||
| GenBank mRNA | AK238319.1 | - | 5 - 6 | 580 | 72 | |
More Information
Set Membership: |
|

LOC100624460
Genomic Map