Assay Details
| Gene Symbol: B3GALT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927568.5 | XP_001927603.1 | 331 | 75 | ||
| GenBank mRNA | AK238427.1 | - | 1 - 2 | 185 | 75 | |
| AK347661.1 | - | 1 - 2 | 185 | 75 | ||
| AK351151.1 | - | 1 - 2 | 138 | 75 | ||
| AK351263.1 | - | 1 - 2 | 190 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001927568.5 transcripts.
Set Membership: |
|

B3GALT4
Genomic Map