Assay Details
| Gene Symbol: ANP32E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021089919.1 | XP_020945578.1 | 590 | 113 | ||
| XM_021089920.1 | XP_020945579.1 | 409 | 113 | |||
| XM_021089921.1 | XP_020945580.1 | 170 | 113 | |||
| XM_021089922.1 | XP_020945581.1 | 531 | 113 | |||
| GenBank mRNA | AK231066.1 | - | 2 - 3 | 508 | 113 | |
| AK343979.1 | - | 2 - 3 | 508 | 113 | ||
| AY609610.1 | - | 2 - 3 | 408 | 113 | ||
More Information
Set Membership: |
|

ANP32E
Genomic Map