Assay Details
| Gene Symbol: LOC100520753 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003128902.4 | XP_003128950.2 | 750 | 109 | ||
| XM_013978575.2 | XP_013834029.1 | 750 | 109 | |||
| GenBank mRNA | AK231070.1 | - | 2 - 3 | 663 | 109 | |
| AK391395.1 | - | 2 - 3 | 724 | 109 | ||
| AK391532.1 | - | 3 - 4 | 575 | 109 | ||
More Information
Set Membership: |
|

LOC100520753
Genomic Map