Assay Details
| Gene Symbol: CLCA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001926978.5 | XP_001927013.4 | 2210 | 86 | ||
| GenBank mRNA | AK231160.1 | - | 5 - 6 | 837 | 86 | |
| AK345076.1 | - | 8 - 9 | 1404 | 86 | ||
| AK345186.1 | - | 8 - 9 | 1403 | 86 | ||
More Information
Set Membership: |
|

CLCA4
Genomic Map