Assay Details
| Gene Symbol: PDZK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005663535.3 | XP_005663592.1 | 1141 | 88 | ||
| XM_013997218.1 | XP_013852672.1 | 932 | 88 | |||
| XM_021089948.1 | XP_020945607.1 | 989 | 88 | |||
| XM_021089949.1 | XP_020945608.1 | 1179 | 88 | |||
| GenBank mRNA | AK231176.1 | - | 6 - 7 | 1189 | 88 | |
| AK394065.1 | - | 5 - 6 | 883 | 88 | ||
| AY609987.1 | - | 6 - 7 | 980 | 88 | ||
More Information
Set Membership: |
|

PDZK1
Genomic Map