Assay Details
| Gene Symbol: GNA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003357998.4 | XP_003358046.1 | 611 | 63 | ||
| GenBank mRNA | AK231228.1 | - | 1 - 2 | 373 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: GNA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003357998.4 | XP_003358046.1 | 611 | 63 | ||
| GenBank mRNA | AK231228.1 | - | 1 - 2 | 373 | 63 | |
Set Membership: |
|