Assay Details
| Gene Symbol: GMPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928014.5 | XP_001928049.1 | 533 | 78 | ||
| XM_021099209.1 | XP_020954868.1 | 123 | 78 | |||
| XM_021099212.1 | XP_020954871.1 | 216 | 78 | |||
| GenBank mRNA | AK231280.1 | - | 2 - 3 | 320 | 78 | |
| AK238993.1 | - | 2 - 3 | 292 | 78 | ||
| AK351787.1 | - | 2 - 3 | 271 | 78 | ||
| AK391336.1 | - | 2 - 3 | 244 | 78 | ||
| AK391520.1 | - | 2 - 3 | 299 | 78 | ||
| AY609855.1 | - | 2 - 3 | 251 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021099212.1 transcripts.
Set Membership: |
|

GMPR2
Genomic Map