Assay Details
| Gene Symbol: MTG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927648.6 | XP_001927683.3 | 349 | 63 | ||
| XM_003483583.4 | XP_003483631.1 | 356 | 63 | |||
| XM_005671585.3 | XP_005671642.1 | 349 | 63 | |||
| XM_005671587.3 | XP_005671644.1 | 431 | 63 | |||
| XM_005671588.3 | XP_005671645.1 | 313 | 63 | |||
| XM_021073959.1 | XP_020929618.1 | 348 | 63 | |||
| XM_021073960.1 | XP_020929619.1 | 347 | 63 | |||
| XM_021073961.1 | XP_020929620.1 | 343 | 63 | |||
| XM_021073962.1 | XP_020929621.1 | 387 | 63 | |||
| XM_021073963.1 | XP_020929622.1 | 431 | 63 | |||
| XM_021073964.1 | XP_020929623.1 | 417 | 63 | |||
| XM_021073965.1 | XP_020929624.1 | 318 | 63 | |||
| GenBank mRNA | AK231290.1 | - | 3 - 4 | 274 | 63 | |
| AK351366.1 | - | 3 - 4 | 291 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005671587.3, XM_005671588.3, XM_021073962.1, XM_021073963.1, XM_021073964.1, XM_021073965.1 transcripts.
Set Membership: |
|

MTG1
Genomic Map