Assay Details
| Gene Symbol: STK17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021074823.1 | XP_020930482.1 | 451 | 133 | ||
| GenBank mRNA | AK231304.1 | - | 2 - 3 | 417 | 133 | |
| HM594868.1 | - | 2 - 3 | 402 | 133 | ||
More Information
Set Membership: |
|

STK17B
Genomic Map