Assay Details
| Gene Symbol: AOPEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021064602.1 | XP_020920261.1 | 2064 | 76 | ||
| XM_021064603.1 | XP_020920262.1 | 1983 | 76 | |||
| XM_021064604.1 | XP_020920263.1 | 2064 | 76 | |||
| XM_021064605.1 | XP_020920264.1 | 2064 | 76 | |||
| GenBank mRNA | AK231317.1 | - | 5 - 6 | 775 | 76 | |
More Information
Set Membership: |
|

AOPEP
Genomic Map