Assay Details
| Gene Symbol: MED10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134157.4 | XP_003134205.1 | 208 | 78 | ||
| XM_013982820.2 | XP_013838274.1 | 211 | 78 | |||
| GenBank mRNA | AK231321.1 | - | 1 - 2 | 133 | 78 | |
| AK346844.1 | - | 1 - 2 | 169 | 78 | ||
| AY610178.1 | - | 1 - 2 | 158 | 78 | ||
More Information
Set Membership: |
|

MED10
Genomic Map