Assay Details
| Gene Symbol: RARRES1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358645.3 | XP_003358693.1 | 699 | 82 | ||
| GenBank mRNA | AK231335.1 | - | 1 - 2 | 317 | 82 | |
| AK235788.1 | - | 1 - 2 | 502 | 82 | ||
| AK395721.1 | - | 1 - 2 | 511 | 82 | ||
| AK399137.1 | - | 1 - 2 | 436 | 82 | ||
More Information
Set Membership: |
|

RARRES1
Genomic Map