Assay Details
Gene Symbol: YAE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003360217.5 | XP_003360265.3 | 315 | 80 | ||
GenBank mRNA | AK231416.1 | - | 2 - 3 | 300 | 80 | |
AK348668.1 | - | 2 - 3 | 283 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
