Assay Details
Gene Symbol: DNAJB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021101721.1 | XP_020957380.1 | 1067 | 76 | ||
XM_021101722.1 | XP_020957381.1 | 878 | 76 | |||
XM_021101724.1 | XP_020957383.1 | 525 | 76 | |||
GenBank mRNA | AK231435.1 | - | 5 - 6 | 763 | 76 | |
AK345305.1 | - | 5 - 6 | 749 | 76 | ||
AK389354.1 | - | 5 - 6 | 870 | 76 | ||
AY609702.1 | - | 2 - 3 | 190 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
