Assay Details
Gene Symbol: C2H11orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003480654.4 | XP_003480702.1 | 546 | 103 | ||
XM_013994015.2 | XP_013849469.1 | 967 | 103 | |||
XM_021082724.1 | XP_020938383.1 | 793 | 103 | |||
GenBank mRNA | AK231440.1 | - | 3 - 4 | 484 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
