Assay Details
| Gene Symbol: CBLIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122682.3 | XP_003122730.2 | 467 | 110 | ||
| GenBank mRNA | AK231492.1 | - | 3 - 4 | 426 | 110 | |
| AK400404.1 | - | 3 - 4 | 426 | 110 | ||
More Information
Set Membership: |
|

CBLIF
Genomic Map