Assay Details
| Gene Symbol: DENR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928848.6 | XP_001928883.1 | 305 | 96 | ||
| GenBank mRNA | AK231522.1 | - | 3 - 4 | 252 | 96 | |
| AK235861.1 | - | 583 | 96 | |||
| AK346540.1 | - | 3 - 4 | 229 | 96 | ||
| AK392720.1 | - | 3 - 4 | 213 | 96 | ||
| AY609734.1 | - | 3 - 4 | 218 | 96 | ||
More Information
Set Membership: |
|

DENR
Genomic Map