Assay Details
| Gene Symbol: MMD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124274.5 | XP_003124322.2 | 282 | 83 | ||
| XM_021086029.1 | XP_020941688.1 | 270 | 83 | |||
| XM_021086030.1 | XP_020941689.1 | 844 | 83 | |||
| XM_021086031.1 | XP_020941690.1 | 422 | 83 | |||
| XM_021086033.1 | XP_020941692.1 | 588 | 83 | |||
| XM_021086034.1 | XP_020941693.1 | 2093 | 83 | |||
| XM_021086035.1 | XP_020941694.1 | 228 | 83 | |||
| XM_021086036.1 | XP_020941695.1 | 281 | 83 | |||
| GenBank mRNA | AK231536.1 | - | 2 - 3 | 421 | 83 | |
More Information
Set Membership: |
|

MMD2
Genomic Map