Assay Details
| Gene Symbol: PLPP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001924179.5 | XP_001924214.1 | 712 | 86 | ||
| GenBank mRNA | AK238542.1 | - | 6 - 7 | 716 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PLPP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001924179.5 | XP_001924214.1 | 712 | 86 | ||
| GenBank mRNA | AK238542.1 | - | 6 - 7 | 716 | 86 | |
Set Membership: |
|