Assay Details
| Gene Symbol: ACSF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131573.4 | XP_003131621.3 | 480 | 97 | ||
| XM_021067162.1 | XP_020922821.1 | 333 | 97 | |||
| GenBank mRNA | AK238561.1 | - | 2 - 3 | 333 | 97 | |
| AK348510.1 | - | 2 - 3 | 330 | 97 | ||
| AK395435.1 | - | 2 - 3 | 485 | 97 | ||
| AK397080.1 | - | 2 - 3 | 428 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021067162.1 transcripts.
Set Membership: |
|

ACSF2
Genomic Map