Assay Details
| Gene Symbol: RRAGC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003127801.4 | XP_003127849.1 | 362 | 61 | ||
| GenBank mRNA | AK238610.1 | - | 1 - 2 | 330 | 61 | |
| AK350262.1 | - | 1 - 2 | 330 | 61 | ||
More Information
Set Membership: |
|

RRAGC
Genomic Map