Assay Details
| Gene Symbol: LSM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003357006.5 | XP_003357054.1 | 2317 | 125 | ||
| XM_005666841.3 | XP_005666898.1 | 2399 | 125 | |||
| XM_005666843.3 | XP_005666900.1 | 2383 | 125 | |||
| XM_005666844.3 | XP_005666901.1 | 2458 | 125 | |||
| XM_005666846.3 | XP_005666903.1 | 2354 | 125 | |||
| XM_013978860.2 | XP_013834314.1 | 2439 | 125 | |||
| XM_013978861.2 | XP_013834315.1 | 2359 | 125 | |||
| XM_021101534.1 | XP_020957193.1 | 2277 | 125 | |||
| GenBank mRNA | AK238655.1 | - | 1 | 113 | 125 | |
More Information
Set Membership: |
|

LSM6
Genomic Map