Assay Details
| Gene Symbol: PPIE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003127817.5 | XP_003127865.1 | 188 | 83 | ||
| XM_005665255.2 | XP_005665312.1 | 375 | 83 | |||
| XM_005665257.2 | XP_005665314.2 | 215 | 83 | |||
| XM_005665258.2 | XP_005665315.1 | 266 | 83 | |||
| XM_013999195.2 | XP_013854649.1 | 374 | 83 | |||
| XM_013999197.2 | XP_013854651.1 | 257 | 83 | |||
| XM_013999198.2 | XP_013854652.1 | 188 | 83 | |||
| XM_013999199.2 | XP_013854653.2 | 215 | 83 | |||
| XM_013999200.2 | XP_013854654.1 | 232 | 83 | |||
| XM_021095990.1 | XP_020951649.1 | 548 | 83 | |||
| GenBank mRNA | AK238670.1 | - | 2 - 3 | 375 | 83 | |
| AK239663.1 | - | 2 - 3 | 142 | 83 | ||
| AK391749.1 | - | 2 - 3 | 141 | 83 | ||
| AK397929.1 | - | 2 - 3 | 134 | 83 | ||
| AK400400.1 | - | 2 - 3 | 150 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005665258.2, XM_013999200.2 transcripts.
Set Membership: |
|

PPIE
Genomic Map