Assay Details
| Gene Symbol: LXN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358644.4 | XP_003358692.1 | 630 | 79 | ||
| GenBank mRNA | AK238706.1 | - | 3 - 4 | 553 | 79 | |
| AK346816.1 | - | 3 - 4 | 552 | 79 | ||
| AK398073.1 | - | 3 - 4 | 571 | 79 | ||
More Information
Set Membership: |
|

LXN
Genomic Map