Assay Details
| Gene Symbol: RETSAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124944.4 | XP_003124992.1 | 1904 | 67 | ||
| GenBank mRNA | AK238789.1 | - | 10 - 11 | 1790 | 67 | |
More Information
Set Membership: |
|

RETSAT
Genomic Map