Assay Details
| Gene Symbol: GJB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021065430.1 | XP_020921089.1 | 2085 | 96 | ||
| GenBank mRNA | AK238800.1 | - | 1 - 2 | 204 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021065430.1 transcripts.
Set Membership: |
|

GJB2
Genomic Map