Assay Details
| Gene Symbol: PPP4R1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013985491.2 | XP_013840945.2 | 216 | 117 | ||
| XM_013985493.2 | XP_013840947.2 | 216 | 117 | |||
| XM_013985494.2 | XP_013840948.2 | 216 | 117 | |||
| XM_013985500.2 | XP_013840954.2 | 216 | 117 | |||
| XM_021078397.1 | XP_020934056.1 | 216 | 117 | |||
| XM_021078398.1 | XP_020934057.1 | 216 | 117 | |||
| XM_021078399.1 | XP_020934058.1 | 216 | 117 | |||
| XM_021078400.1 | XP_020934059.1 | 216 | 117 | |||
| XM_021078401.1 | XP_020934060.1 | 216 | 117 | |||
| XM_021078404.1 | XP_020934063.1 | 216 | 117 | |||
| XM_021078405.1 | XP_020934064.1 | 216 | 117 | |||
| GenBank mRNA | AK238827.1 | - | 2 - 3 | 187 | 117 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013985491.2, XM_013985493.2, XM_013985494.2, XM_013985500.2, XM_021078397.1, XM_021078398.1, XM_021078399.1, XM_021078400.1, XM_021078401.1, XM_021078404.1, XM_021078405.1 transcripts.
Set Membership: |
|

PPP4R1L
Genomic Map