Assay Details
| Gene Symbol: RPS15A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021086553.1 | XP_020942212.1 | 556 | 75 | ||
| GenBank mRNA | AK238842.1 | - | 1 - 2 | 390 | 75 | |
| AK398844.1 | - | 1 - 2 | 556 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021086553.1 transcripts.
Set Membership: |
|

RPS15A
Genomic Map