Assay Details
| Gene Symbol: SWAP70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122981.6 | XP_003123029.2 | 552 | 60 | ||
| GenBank mRNA | AK238957.1 | - | 1 - 2 | 156 | 60 | |
| AK392943.1 | - | 1 - 2 | 173 | 60 | ||
More Information
Set Membership: |
|

SWAP70
Genomic Map