Assay Details
| Gene Symbol: TLE5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021084158.1 | XP_020939817.1 | 549 | 74 | ||
| GenBank mRNA | AK238961.1 | - | 6 - 7 | 524 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: TLE5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021084158.1 | XP_020939817.1 | 549 | 74 | ||
| GenBank mRNA | AK238961.1 | - | 6 - 7 | 524 | 74 | |
Set Membership: |
|