Assay Details
| Gene Symbol: GET3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003123324.4 | XP_003123372.1 | 501 | 102 | ||
| GenBank mRNA | AK238998.1 | - | 2 - 3 | 337 | 102 | |
More Information
Set Membership: |
|

GET3
Genomic Map