Assay Details
| Gene Symbol: ASXL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003125333.6 | XP_003125381.5 | 7746 | 86 | ||
| XM_021087739.1 | XP_020943398.1 | 7826 | 86 | |||
| XM_021087740.1 | XP_020943399.1 | 7953 | 86 | |||
| XM_021087741.1 | XP_020943400.1 | 8766 | 86 | |||
| GenBank mRNA | AK239003.1 | - | 1 | 1921 | 86 | |
More Information
Set Membership: |
|

ASXL2
Genomic Map