Assay Details
| Gene Symbol: CRKL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003132994.4 | XP_003133042.1 | 3585 | 128 | ||
| GenBank mRNA | AK239106.1 | - | 1 | 520 | 128 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CRKL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003132994.4 | XP_003133042.1 | 3585 | 128 | ||
| GenBank mRNA | AK239106.1 | - | 1 | 520 | 128 | |
Set Membership: |
|