Assay Details
| Gene Symbol: PRSS16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021099809.1 | XP_020955468.1 | 783 | 60 | ||
| GenBank mRNA | AK239111.1 | - | 7 - 8 | 787 | 60 | |
| AK398418.1 | - | 7 - 8 | 787 | 60 | ||
More Information
Set Membership: |
|

PRSS16
Genomic Map