Assay Details
| Gene Symbol: RNF135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131735.4 | XP_003131783.1 | 410 | 61 | ||
| GenBank mRNA | AK239120.1 | - | 1 - 2 | 389 | 61 | |
More Information
Set Membership: |
|

RNF135
Genomic Map