Assay Details
| Gene Symbol: LOC100511937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013999442.2 | XP_013854896.1 | 5749 | 91 | ||
| XM_013999444.2 | XP_013854898.2 | 4858 | 91 | |||
| XM_021096719.1 | XP_020952378.1 | 4649 | 91 | |||
| XM_021096721.1 | XP_020952380.1 | 4648 | 91 | |||
| GenBank mRNA | AK239198.1 | - | 1 | 1216 | 91 | |
| AK349215.1 | - | 1 | 1937 | 91 | ||
More Information
Set Membership: |
|

LOC100511937
Genomic Map