Assay Details
Gene Symbol: C7H6orf89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013977868.2 | XP_013833322.2 | 1388 | 91 | ||
XM_013977869.2 | XP_013833323.2 | 1120 | 91 | |||
XM_013977870.2 | XP_013833324.2 | 1053 | 91 | |||
XM_021098650.1 | XP_020954309.1 | 1564 | 91 | |||
XM_021098651.1 | XP_020954310.1 | 1107 | 91 | |||
XM_021098652.1 | XP_020954311.1 | 1658 | 91 | |||
XM_021098653.1 | XP_020954312.1 | 979 | 91 | |||
GenBank mRNA | AK239396.1 | - | 7 - 8 | 1049 | 91 | |
AK390756.1 | - | 7 - 8 | 1057 | 91 | ||
AK390877.1 | - | 7 - 8 | 1023 | 91 | ||
AK394782.1 | - | 8 - 9 | 1174 | 91 | ||
AK396826.1 | - | 7 - 8 | 993 | 91 | ||
AK398191.1 | - | 7 - 8 | 1019 | 91 | ||
AK400725.1 | - | 7 - 8 | 1021 | 91 | ||
AK400837.1 | - | 7 - 8 | 1045 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
