Assay Details
| Gene Symbol: SRSF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021098645.1 | XP_020954304.1 | 556 | 108 | ||
| XM_021098646.1 | XP_020954305.1 | 601 | 108 | |||
| GenBank mRNA | AK239497.1 | - | 5 - 6 | 584 | 108 | |
| AK390451.1 | - | 5 - 6 | 605 | 108 | ||
| AK393899.1 | - | 4 - 5 | 306 | 108 | ||
More Information
Set Membership: |
|

SRSF3
Genomic Map