Assay Details
| Gene Symbol: USP22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021068045.1 | XP_020923704.1 | 2472 | 73 | ||
| GenBank mRNA | AK239526.1 | - | 1 | 225 | 73 | |
| AK392010.1 | - | 13 | 2445 | 73 | ||
More Information
Set Membership: |
|

USP22
Genomic Map