Assay Details
| Gene Symbol: ANP32B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021066477.1 | XP_020922136.1 | 409 | 69 | ||
| GenBank mRNA | AK239532.1 | - | 1 - 2 | 324 | 69 | |
| AK390985.1 | - | 1 - 2 | 414 | 69 | ||
More Information
Set Membership: |
|

ANP32B
Genomic Map