Assay Details
| Gene Symbol: SKP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021084761.1 | XP_020940420.1 | 769 | 79 | ||
| GenBank mRNA | AK239534.1 | - | 4 - 5 | 424 | 79 | |
| AK391651.1 | - | 4 - 5 | 436 | 79 | ||
| AK392736.1 | - | 4 - 5 | 410 | 79 | ||
| AY610484.1 | - | 3 - 4 | 319 | 79 | ||
More Information
Set Membership: |
|

SKP1
Genomic Map