Assay Details
| Gene Symbol: PSMD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005664584.3 | XP_005664641.1 | 648 | 74 | ||
| GenBank mRNA | AK239689.1 | - | 3 - 4 | 366 | 74 | |
| AK394396.1 | - | 3 - 4 | 377 | 74 | ||
More Information
Set Membership: |
|

PSMD8
Genomic Map